Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Silver nitrate 5% in aqueous solution

Supplier: Electron Microscopy Sciences
Synonyms: Silver mononitrateNitric acid silver saltLunar caustic

26212-01
100504-572EA 142.41 USD InStock
100504-572
Silver nitrate 5% in aqueous solution
Silver nitrate
Pizzolato’s Method for Calcium Oxalate is used for bone and its related stain. Stain results - Calcium oxalate: Black, All other tissue elements: Red.
Solutions to use, Silver Nitrate 5% Aqueous, Hydrogen Peroxide 30%, Nuclear Fast Red (Kernechtrot) Solution
Pizzolato, P., J. Histochem. Cytochem., 12:333-136, 1964.
Luna, L.G., (ed), Manual of Staining Methods of the AFIP, 3rd Ed, McGraw-Hill, NY, c 1968, p 77.


Formula: AgNO₃
MW: 169.87 g/mol
MDL Number: MFCD00003414
CAS Number: 7761-88-8
https://us-legacy.vwr.com/stibo/web/std.lang.all/06/45/16640645.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty