Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium hexacyanoferrate(II) 20% in aqueous solution

Supplier: Electron Microscopy Sciences
Warning

25154-20
101410-856EA 55.53 USD InStock
101410-856
Potassium hexacyanoferrate(II) 20% in aqueous solution
Potassium hexacyanoferrate (II)
  • Potassium Ferrocyanide is available for your convenience in 2%, 5%, 10%, or 20% aqueous solutions.


Formula: C₆FeK₄N₆
MW: 368.35 g/mol
Boiling Pt: 100 °C (212 °F)
Density: 1,178 g/cm³ (9.83 lbs/gal.)
MDL Number: MFCD00167022
CAS Number: 13943-58-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty