Would you like to log in again?
Vydac Denali Silica, HPLC Columns
Supplier: VWR
Vydac Denali
5174066 5174054 5174057 5174063 5174061
75812-962EA 3694.68 USD InStock
75812-962 75812-960 75799-026 75799-028 75812-958
Vydac Denali Silica, HPLC Columns
Chromatography Columns
Vydac Denali (238DE) is a 120 Å C18 bonded phase with high carbon coverage, suitable for the analysis of both acidic and basic analytes
  • High retentivity
  • LC-MS of small molecules
  • Fully scalable from capillary to process
  • USP L3
It has applications for small molecule analyses of interest to pharmaceutical and environmental laboratories.
Vydac is a registered trademark of W.R.Grace and Co.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty