Would you like to log in again?
Avantor® Prevail Phenyl, HPLC Columns
Supplier: VWR
Avantor® Prevail
99237 99130/N 99241 99326 99239 99288/N 99327
75830-696EA 356.78 USD InStock
75830-696 75797-852 75830-708 75797-854 76398-350 75797-858 75797-848
Avantor® Prevail Phenyl, HPLC Columns
Chromatography Columns
Avantor® Hichrom Prevail™ HPLC columns exhibit long column lifetime in both highly aqueous and highly organic mobile phases
  • Stable from highly organic to highly aqueous
  • Specialty phases for specific applications
  • Excellent sensitivity with microbore and ELSD applications
  • USP L11
Selective for aromatic compounds in a variety of mobile phase conditions.
All phases, apart from Avantor® Hichrom Prevail™ Carbohydrate ES are silica-based. The stability of these phases is such that a single column can be switched between highly aqueous for analysis of highly polar analytes and highly organic for strong retention of hydrophobic analytes.
Accessories information: Use guard cartridges with holder (80101/N) and coupler (HI-081).
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty