Would you like to log in again?
SHOWA 7066 Fully Coated Nitrile, General-Purpose Glove, Showa
Supplier: Showa
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
7066-10 7066-09 7066-08
32891-860CS 653.4 USD InStock
32891-860 32891-862 32891-854
SHOWA 7066 Fully Coated Nitrile, General-Purpose Glove, Showa
Gloves
100% Nitrile safety cuff general-purpose glove provides excellent protection in highly abrasive applications.
  • 100% Nitrile
  • Safety cuff
  • Cotton jersey liner
These gloves are designed for tough abrasive applications to replace leather and cotton gloves.
Certifications: ANSI Abrasion Level 4.
Packaging: Packaged 12 pair per bag.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty