Would you like to log in again?
SHOWA 960 PVC General-Purpose Work Glove, Showa
Supplier: Showa
960M-09 960S-08 960L-10
32891-922CS 579.16 USD InStock
32891-922 32891-921 32891-923
SHOWA 960 PVC General-Purpose Work Glove, Showa
Gloves
Seamless PVC coated work gloves are made in the U.S.
  • PVC coating for abrasive applications
  • Breathable coating for comfort
Gloves are designed for use in assembly, grounds/maintenance, and material handling applications.
Certifications: ANSI Abrasion level 3.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty