Would you like to log in again?
Jacobson Micro Needle Holder with Catch, Sklar
Supplier: Sklar
98-3171 98-3170
10185-306EA 848.54 USD InStock
10185-306 10185-308
Jacobson Micro Needle Holder with Catch, Sklar
Forceps
Sklar's® Jacobson Micro Needle Holder is primarily used for suturing in microvascular procedures.
  • Can hold very fine sutures
  • Nonsterile and reusable
The needle holder may also be known as a Jacobson-Castroviejo Needle Holder. The finely cross-serrated, tapered jaws are able to securely hold very fine sutures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty