Would you like to log in again?
ZING Green Safety Green at Work Sign, Glass Only, Recycle Symbol
Supplier: ZING Green Safety Products
1030 2030 3030 1030S 0030S 2030S
76023-392PK 59.32 USD InStock
76023-392 76025-450 76024-400 76022-604 76023-390 76024-402
ZING Green Safety Green at Work Sign, Glass Only, Recycle Symbol
Signs
Adhesive labels are available in three sizes, 7" x 5", 10" x 7", and 14" x 10" with 5/Pack. Triview signs are 1/Pack.
  • UL Validated for recycled content
  • Made in USA
  • High quality
Adhesive signs are made from a minimum of 70% post-industrial polystyrene, and are resistant to solvents, moisture, scuffing, tears, and UV exposure
Three-sided Tri-View Sign offers a 180-degree view giving improved visibility. Sign is made from high quality 70% post-consumer recycled plastic. Sign includes pre-drilled holes and self-adhesive pads for easy mounting and measures 7-1/2"H x 20"W and projects 5 inches. Made in the USA.
The signs adhere directly to walls, recycling bins, and waste receptacles. All Zing signs are made in the USA from recycled materials, which supports sustainability initiatives and LEED credits. The signs are sold 5/pack.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty