Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
ZING Green Safety Eco Safety Tag, DANGER Do Not Operate
Supplier: ZING Green Safety Products
7010
76026-012PK 46.8 USD InStock
76026-012
ZING Green Safety Eco Safety Tag, DANGER Do Not Operate
Signs
Communicates critical safety information regarding locked out equipment.
  • High quality 70% post-consumer recycled plastic with a surface that is easy to write on
  • Made in USA
  • Exceeds OSHA 50 pound pull test
ZING Eco Safety Lockout Tags "DANGER DO NOT OPERATE" communicate critical safety information regarding locked out equipment. Tags are made from rigid .055 gauge high quality 70% post-consumer recycled plastic with a surface that is easy to write on. Durable tags hold up against water, grease and a variety of chemicals making them suitable for indoor or outdoor use. Recycled material is UL Validated for recycled content which substantiates material compliance with various green codes, standards, and procurement policies to support sustainability initiatives and LEED credits. Tags measure 5.75"H x 3"W and hole measures 1/2". Exceeds OSHA's 50 lb. pull requirement. 10 Tags per Pack. Made in the USA.
Certifications: 29 CFR 1910.147
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty