Would you like to log in again?
Economy Tesla Meter
Supplier: OSAW INDUSTRIAL PRODUCTS SE
SE1093
470229-042EA 704.9 USD InStock
470229-042
Economy Tesla Meter
Data Loggers
Measure magnetic fields quickly and accurately.
  • Suitable for measuring magnetic fields up to 2 T
  • Adjustment knob to set the magnetic field to zero
Ideal for tangential direct magnetic field measurements for hall effect and other magnetic field based labs. Two switchable ranges and a three digit LED display.
Range measurements are:
0 to 200 mT at a resolution of 0.1 mT
0 to 1999 mT at a resolution of 1 mT
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty