Would you like to log in again?
Radioactive Bundle
Supplier: SCIENCE INTERACTIVE GROUP
615-1500 615-1510
470229-066KT 1593.39 USD InStock
470229-066 470229-068
Radioactive Bundle
Educational Materials Physics Educational Materials Modern Physics Learning Activities
A complete modern physics unit in one bundle.
  • Includes instruction manual
  • Alpha, Beta, and Gamma source
This bundle includes 8 lessons and 3 teacher demonstrations. Contents include:
Millikan Apparatus
Half-life calculations worksheet
Brazil nut radioactivity/half-life experiment
Calculating total radiation exposure based on a food’s mass
Carbon-14 dating graphing exercise
Build a particle detector
Cloud Chamber Variables
Radioactivity in My Classroom Environment
Research project on medical applications of radioactive imaging
The bundle also includes a cloud chamber, radioactive sources (Alpha, Beta, and Gamma), Millikan apparatus, and Geiger counter. The deluxe set also includes an e/m apparatus.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty