Would you like to log in again?
SHOWA S8115 HPPE Cut Resistant Sleeve, 16 Length, Showa
Supplier: Showa
S8115M-16 S8115L-16
89208-346CS 519.98 USD InStock
89208-346 89208-344
SHOWA S8115 HPPE Cut Resistant Sleeve, 16 Length, Showa
Workwear
HPPE material provides excellent comfort and cut resistance.
  • HPPE cut resistant material (ANSI A4)
  • 16" length
  • Launderable and bleachable
Sleeves are designed to protect arms from cuts. The ability to launder and bleach is a plus in many environments.
Sleeve can be easily laundered.
Certifications: ANSI A4 Cut Level.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty