Would you like to log in again?
Whatman™ Grade 226 Qualitative Filter Papers, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
Whatman™
5226-190 5226-200 5226-330 5226-200 5226-330 5226-190 5226-848 5226-848
28297-614PK 107.05 USD InStock
CA28297-614 28297-614 CA28325-130 28325-103 28325-125 CA28325-125 CA28325-103 28325-130
Whatman™ Grade 226 Qualitative Filter Papers, Whatman products (Cytiva)
Filters Filter Paper Qualitative Filter Paper
A general qualitative filter with creped surface. Widely used in industrial or student laboratories for routine filtration.
  • High filtration speed
  • Retains coarse or gelatinous particulates
  • Unique creped gray surface provides optical contrast for white or dark filtrates
The filter is wet-strengthened with a small amount of chemically stable resin to extend the life of the filter and improve handling.
Designed for routine and repetitive applications for which an inexpensive product is acceptable, such as qualitative analysis.
Normal qualitative applications will not introduce any significant impurities into the filtrate. However it is not recommended for Kjeldahl nitrogen analysis.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty