Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium borohydride, crystalline powder ACS

Supplier: SPECTRUM LABORATORY PRODS LO
Danger
Synonyms: Sodium borohydride Sodium tetrahydridoborate

S1187-10
80701-854EA 1484.44 USD InStock
80701-854
Sodium borohydride, crystalline powder ACS
Sodium borohydride


Formula: NaBH₄
MW: 37.83 g/mol
MDL Number: MFCD00003518
CAS Number: 16940-66-2
UN: 1426
ADR: 4.3,I
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty