Would you like to log in again?
SP Wilmad-LabGlass Oil Centrifuge Tube with Short Stem, SP Industries
Supplier: SP Industries
LG-4420-100
14202-752EA 487.45 USD InStock
14202-752
SP Wilmad-LabGlass Oil Centrifuge Tube with Short Stem, SP Industries
Tubes Centrifuge Tubes
This Goetz-type oil centrifuge tube with short stem was originally designed for determination of bottom sediment and water in petroleum and its products.
Made of borosilicate glass, it is calibrated 'to contain' (TC). Its' approximate widest Ext.Ø is 58 mm, and the approximate length is 158 mm.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty