Would you like to log in again?
Casein-peptone Lecithin Polysorbate Broth Base Dehydrated Culture Media, MilliporeSigma
Supplier: MilliporeSigma
1.11723.0500 1.11723.0500
EM1.11723.0500EA 268.85 USD InStock
CA1.11723.0500 EM1.11723.0500
Casein-peptone Lecithin Polysorbate Broth Base Dehydrated Culture Media, MilliporeSigma
Media for Microbiology Dehydrated Media for Microbiology
Media is granulated.
  • Granulation preserves homogeneity
  • Easy to weigh out
  • Dissolves faster than powdered media
Granulation preserves homogeneity by capturing the media’s original component distribution in each granule and preventing settling out. Dangers associated with inhalation of fine dust and toxic substances are significantly reduced with granulated media. It is easier to weigh out and dissolves faster than powdered media.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty