Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ubiquitin Activating Kit, Enzo Life Sciences

Supplier: Enzo Life Sciences
UW0400A-0001
76002-186EA 261.66 USD InStock
76002-186
Ubiquitin Activating Kit, Enzo Life Sciences
Assays Enzyme Assays
Uses:

  • For use with wild type or mutant E2 enzymes in E1 initiated/mediated reactions
  • Contains highly sensitive ubiquitin antibody for detection of ubiquitin and ubiquitin conjugates

For activation of ubiquitin for use in ubiquitinylation experiments. The kit provides the means of generating thioester linked, activated ubiquitin-E1 conjugates, utilizing the first step in the ubiquitin cascade, for investigation of ubiquitin activation, subsequent ubiquitin transfer to/interaction with E2 conjugating enzymes, and their use in the ubiquitinylation of E3 ligases and target substrate proteins. Ubiquitin antibody provided allows high sensitivity detection of ubiquitin and ubiquitin-conjugates.

Ordering information: Contains 20X Ubiquitin Activating Enzyme Solution, Ubiquitin Solution, Ubiquitin Antibody Solution, Ubiquitin protein conjugates rabbit PAb, 20X Mg-ATP Solution, 2X Non-reducing Gel Loading Buffer, 10X Ubiquitinylation Buffer.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty