Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Oscilloscope Probe Set

Supplier: CAL TEST ELECTRONICS SE
CT2704
470123-088EA 32.65 USD InStock
470123-088
Oscilloscope Probe Set
Electrical Meters Digital Multimeters
Excellent replacement probes for all makes and models of oscilloscopes.

  • Snap-locking sprung hook
  • Easily replaceable tip
  • Large accessory set
  • 100 MHz (-3 dB) bandwidth
  • High impedance input (10 MΩ/16 pF - 10x position)
  • 10−35 pF compensation range
  • 300 V CAT II

General purpose series 100 MHz, 1x/10x switchable oscilloscope probe.

The High Performance family is a high quality, general purpose set of 5 mm Passive Voltage Oscilloscope probes.

Certifications: Meets IEC 61010-031. RoHS compliant.

Ordering information: Accessories included are 15 cm Ground Lead, Replacement Tip, Black Snap-on Sprung Hook, Tip Insulator, IC Tip, BNC Adapter, Probe Tip Ground, Trimmer Tool and Identifier Rings.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty