Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Galloflavin ≥95% (by HPLC)

Supplier: BioVision
Synonyms: NSC-1070223,8,9,10-Tetrahydroxy-pyrano[3,2-c][2]benzopyran-2,6-dione

AB141776-25MG AB141776-5MG
76021-050EA 1264.05 USD InStock
76021-050 76021-048
Galloflavin ≥95% (by HPLC)
Galloflavin
Galloflavin is a lactate dehydrogenase (LDH) inhibitor inhibiting both LDH-A and LDH-B isoforms of the enzyme with Ki values of 5.46 a µM and 15.06 µM, respectively. It blocks the proliferation of cancer cells by blocking glycolysis and ATP production.

  • Formulation: Light yellow to light brown powder


Formula: C₁₂H₆O₈
MW: 278.17 g/mol
Storage Temperature: Freezer
MDL Number: MFCD26941292
CAS Number: 568-80-9
https://us-legacy.vwr.com/stibo/web/std.lang.all/21/99/23492199.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty