Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

GSK429286A ≥98% (by HPLC)

Supplier: BioVision
Synonyms: RHO-15GSK429286N-(6-Fluoro-1H-indazol-5-yl)-2-methyl-6-oxo-4-(4-(trifluoromethyl)phenyl)-1,4,5,6-tetrahydropyridine-3-carboxamide4-[4-(Trifluoromethyl)phenyl]-N-(6-Fluoro-1H-indazol-5-yl)-2-methyl-6-oxo-1,4,5,6-tetrahydro-3-pyridinecarboxamide

AB146581-1MG AB146581-5MG
76021-318EA 221.82 USD InStock
76021-318 76021-320
GSK429286A ≥98% (by HPLC)
GSK429286A
GSK429286A is a potent and selective Rho-Kinase (ROCK) inhibitor (IC₅₀ = 14 and 63 nM for ROCK1 and ROCK2, respectively). ROCK inhibitors have been found to prevent apoptosis, as well as enhance the survival and cloning efficiency of dissociated human embryonic stem (ES) cells without affecting their self-renewal properties or pluripotency.

  • Appearance: White solid


Formula: C₂₁H₁₆F₄N₄O₂
MW: 432.4 g/mol
Storage Temperature: Freezer
MDL Number: MFCD16618404
CAS Number: 864082-47-3
https://us-legacy.vwr.com/stibo/web/std.lang.all/21/65/23492165.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty