Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

GSK429286A , EZSolution™

Supplier: BioVision
Synonyms: RHO-15GSK429286N-(6-Fluoro-1H-indazol-5-yl)-2-methyl-6-oxo-4-(4-(trifluoromethyl)phenyl)-1,4,5,6-tetrahydropyridine-3-carboxamide4-[4-(Trifluoromethyl)phenyl]-N-(6-Fluoro-1H-indazol-5-yl)-2-methyl-6-oxo-1,4,5,6-tetrahydro-3-pyridinecarboxamide

AB146632-1MG
76021-326EA 268.13 USD InStock
76021-326
GSK429286A , EZSolution™
GSK429286A
A 10 mM (1 mg in 231 µl) of the ROCK inhibitor GSK429286A (Cat

No. 2637) in anhydrous DMSO.


Formula: C₂₁H₁₆F₄N₄O₂
MW: 432.4 g/mol
Storage Temperature: Freezer
MDL Number: MFCD16618404
CAS Number: 864082-47-3
https://us-legacy.vwr.com/stibo/web/std.lang.all/21/68/23492168.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty