Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

DPQ , EZSolution™

Supplier: BioVision

AB146611-5MG
76021-840EA 720.15 USD InStock
76021-840
DPQ , EZSolution™
DPQ
A 10 mM (5 mg in 1653 µl) of the PARP inhibitor DPQ (Cat

No. 1899) in anhydrous DMSO.


Formula: C₁₈H₂₆N₂O₂
MW: 302.4
Storage Temperature: Freezer
MDL Number: MFCD01866274
CAS Number: 129075-73-6
https://us-legacy.vwr.com/stibo/web/std.lang.all/23/40/23492340.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty