Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Aftin-5 ≥98% (by NMR)

Warning
Synonyms: (2R)-2-[[9-Isopropyl-6-(N-methylanilino)purin-2-yl]amino]butan-1-olAmyloidβ Forty-Two Inducer 5

AB285376-1MG AB285376-5MG
76021-096EA 206.84 USD InStock
76021-096 76021-098
Aftin-5 ≥98% (by NMR)
Aftin-5
Roscovitine-related purine with no activity on CDKS.

Selectively and potently increases production of extracellular Aβ42 and decreases production of extracellular Aβ38 in cultured cells. Extracellular Aβ40 levels remain stable. Intracellular levels of these amyloids appear to remain stable. Alzheimer's Disease (AD) accelerator that interacts with VDAC1, prohibitin and mitofilin, possibly interfering with subcellular compartmentalization and lipid rafts properties, shifting γ-secretase activity toward Aβ42 generation. Induces a reversible mitochondrial phenotype reminiscent of the one observed in AD brains.

Storage Temperature: -20°C


Formula: C19H26N6O
MW: 354.5 g/mol
CAS Number: 1461717-05-4
https://us-legacy.vwr.com/stibo/web/std.lang.all/21/00/23492100.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty