Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pluronic® F-127 (Poloxamer 407), M.W. ~12500

Supplier: BioVision
Synonyms: Polyethylene-Polypropylene Glycol

AB285413-50G AB285413-250G
76021-574EA 199.04 USD InStock
76021-574 76021-576
Pluronic® F-127 (Poloxamer 407), M.W. ~12500
Pluronic® F-127 (Poloxamer 407)
Pluronic® F-127 is a non-ionic detergent useful for isolation of membrane proteins. It has been found to facilitate the solubilization of water-insoluble dyes and other materials in physiological media.

Storage Temperature: RT


Formula: HO(C2H4O)a(C3H6O)b(C2H4O)aH
MDL Number: MFCD00082049
CAS Number: 9003-11-6
https://us-legacy.vwr.com/stibo/web/std.lang.all/22/48/23492248.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty