Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pluronic® F-127 (Poloxamer 407) 10% in aqueous solution, MegPure™, Proteomics Grade, sterile filtered

Supplier: BioVision
Synonyms: Polyethylene-Polypropylene Glycol

AB288152-500ML AB288152-100ML AB288152-25ML AB288152-50ML
76021-578EA 164.54 USD InStock
76021-578 76021-580 76021-582 76021-584
Pluronic® F-127 (Poloxamer 407) 10% in aqueous solution, MegPure™, Proteomics Grade, sterile filtered
Pluronic® F-127 (Poloxamer 407)
Pluronic® F-127 is a non-ionic detergent useful for isolation of membrane proteins. It has been found to facilitate the solubilization of water-insoluble dyes and other materials in physiological media.

Storage Temperature: +4°C


Formula: HO(C2H4O)a(C3H6O)b(C2H4O)aH
MDL Number: MFCD00082049
CAS Number: 9003-11-6
https://us-legacy.vwr.com/stibo/web/std.lang.all/22/49/23492249.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty