Would you like to log in again?
Multipurpose Mini Spin Columns, Biovision Inc.
Supplier: BioVision
AB286861-50UNITS
10835-324EA 296.35 USD InStock
10835-324
Multipurpose Mini Spin Columns, Biovision Inc.
Filters Centrifugal Filters
Columns aiding in IP, antibody purification, protein desalting etc.
  • Constructed of polypropylene with a polyethylene FRIT
  • Porosity of 20 µm and a total bed volume of 700 µl
Protein activity and functional studies rely largely on the availability of purified proteins. Multipurpose Mini Spin Columns offer a convenient option to purify target proteins by multiple small scale laboratory techniques. Along with affinity purification and recombinant protein purification, these columns can also be used for techniques like immunoprecipitation, desalting etc.
Each of these columns can be packed with a variety of media/beads for centrifugation-based separation of biomolecules. The columns are equipped with tight top screw caps and a bottom plugs making them ideal also for storage of beads/media in buffers, incubation of beads with biological samples, etc.
Selected Applications:
Immunoprecipitation and co-immunoprecipitation with protein A/G beads (10008-748, 10008-844, 10008-876, 10008-888, 10008-902)
Antibody purification using protein A, G, L beads ((10008-748, 10008-844, 10008-876, 10008-888, 10008-902, 10008-934, 10008-950)
Affinity protein purification with beads like the EZEnrich™ Polyubiquitin Beads (10835-320)
Protein/DNA desalting using Sephadex G25 or Sephadex G50
Recombinant protein affinity purification of proteins with polyhistidine tag, GST tag, etc.
Caution: For research use only. Not for use in humans.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty