Would you like to log in again?
ApoSENSOR™ ATP Cell Viability Bioluminescence Assay Kit, BioVision
Supplier: BioVision
AB65314-200
10013-664EA 259.62 USD InStock
10013-664
ApoSENSOR™ ATP Cell Viability Bioluminescence Assay Kit, BioVision
Assays Cellular Assays
Ultra-sensitive Assay, HTS
  • Simple one-step procedure; takes only 30 minutes
  • Fast and convenient
  • The assay can be done directly in culture plates requiring no harvest/washing/or sample preparations. The assay can be fully automatic for high throughput (10 seconds/sample) and is highly sensitive (detects 10-100 mammalian cells/well).
Cell death (especially apoptosis) is an energy-dependent process that requires ATP. As ATP levels fall to a point where the cell can no longer perform basic metabolic functions, the cell will die. A typical apoptotic cell exhibits a significant decrease in ATP level. Therefore, loss of ATP level in cell has been used as an indicator of cell death. In contrast, cell proliferation has been recognized by increased levels of ATP. The ApoSENSOR™ Cell Viability Assay Kit utilizes bioluminescent detection of the ATP levels for a rapid screening of apoptosis and cell proliferation simultaneously in mammalian cells. The assay utilizes luciferase to catalyze the formation of light from ATP and luciferin, and the light can be measured using a luminometer or Beta Counter. The assay can be fully automatic for high throughput (10 seconds/sample) and is extremely sensitive (detects 10-100 mammalian cells/well). The high sensitivity of this assay has led to many other applications for detecting ATP production in various enzymatic reactions, as well as for detecting low level bacterial contamination in samples such as blood, milk, urine, soil, and sludge.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty