Would you like to log in again?
Glycerol-3-Phosphate Dehydrogenase Activity Colorimetric Assay Kit, BioVision Inc.
Supplier: BioVision
AB174095-100TEST
10013-766EA 1093.83 USD InStock
10013-766
Glycerol-3-Phosphate Dehydrogenase Activity Colorimetric Assay Kit, BioVision Inc.
Assays Enzyme Assays
Highly-sensitive assay to measure GPDH activity
  • Simple, sensitive, and rapid.
  • The assay can detect Glycerol-3-Phosphate dehydrogenase activity less than 1 mU/well.
Glycerol-3-Phosphate Dehydrogenase (EC 1.1.1.8) is an important enzyme for lipid metabolism. It catalyzes the reversible conversion between dihydroxyacetone phosphate and glycerol-3-phosphate. GPDH plays multiple functions inside cells; it links carbohydrate and lipid metabolism, and provides electrons through the Glycerol-3-Phosphate Shuttle. When progenitor adipocytes differentiate into mature adipocytes, GPDH activity increases significantly. Analysis of glycerol-3-phosphate dehydrogenase activity is crucial for the study of fatty acid metabolic pathways. In BioVision’s Glycerol-3-Phosphate Dehydrogenase Activity Assay kit, Glycerol-3-phosphate dehydrogenase forms an intermediate, which reduces a colorless probe to a colored product with strong absorbance at 450 nm. The assay is simple, sensitive and rapid and can detect Glycerol-3-Phosphate dehydrogenase activity less than 1 mU/well.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty