Would you like to log in again?
Enteropeptidase/Enterokinase Inhibitor Screening Kit (Fluorometric), BioVision
Supplier: BioVision
AB170965-100TEST
10013-788EA 1149.14 USD InStock
10013-788
Enteropeptidase/Enterokinase Inhibitor Screening Kit (Fluorometric), BioVision
Assays Enzyme Assays
Simple Assay to Screen Enteropeptidase Inhibitors
  • Rapid & simple
  • Reliable test for screening potential inhibitors of Enteropeptidase/Enterokinase
  • Includes Enteropeptidase-specific Inhibitor [Aprotinin]
Enteropeptidase (Enterokinase, EC 3.4.21.9) is a serine protease involved in activation of trypsinogen to trypsin, which in turn results in the activation of various digestive enzymes. It recognizes a highly specific amino acid sequence ‘DDDDK’ and cleaves after the lysine residue. High specific activity of Enteropeptidase has been utilized in cleaving a variety of native or fusion proteins containing the above recognition. In Biovision’s Enteropeptidase inhibitor screening kit, we have utilized a peptide substrate containing the Enteropeptidase recognition sequence along with a fluorescent label ‘AFC’. Enteropeptidase catalyzes the cleavage of this substrate and releases the AFC molecule, which can be easily quantified by measuring its fluorescence at Ex/Em = 380/500 nm. In the presence of potent Enteropeptidase inhibitor, the hydrolyzation of the substrate will be impeded. The kit provides a rapid, simple & reliable test for screening potential inhibitors of Enteropeptidase.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty