Would you like to log in again?
EZCellTM Cell Invasion Assay (Basement Membrane), 96-well, 8 µm, BioVision
Supplier: BioVision
AB235697-100TEST
10013-828EA 982.37 USD InStock
10013-828
EZCellTM Cell Invasion Assay (Basement Membrane), 96-well, 8 µm, BioVision
Assays Cellular Assays
Highly Sensitive & Quantitative (Fluorometric) Assay to measure Cell Invasion using Boyden Chamber
  • Highly sensitive fluorometric method to measure cell invasion in response to a variety of biochemical stimuli
  • Simple & High throughput-adaptable
  • Reproducible, Quantitative tool for screening, studying, and characterizing compounds that affect cell invasion
Cell invasion is the ability of cells to migrate from one area to another through an extracellular matrix. Cell invasion is exhibited by both normal cells as well as cancerous cells in response to specific external signals, including chemical & mechanical stimuli. During invasion, extracellular matrix is enzymatically degraded by cellular proteases before cells migrate to the new location. Cell invasion is required for normal processes such as wound repair, vasculature formation and the inflammatory response as well as the abnormal invasion of tissues by tumor cells during metastasis. BioVision’s Cell Invasion Assay Kit utilizes a Boyden chamber coated with Basement Membrane Extract (BME), where the cells invade the matrix and then migrate through a semipermeable membrane in the Boyden chamber in response to stimulants or inhibitory compounds. The percent cell invasion can be analyzed directly in a plate reader. Our assay is easy to use, sensitive and adaptable to high-throughput systems.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty