Would you like to log in again?
Oxalate (Oxalic Acid) Colorimetric Assay Kit, BioVision
Supplier: BioVision
AB196990-100TEST
10191-110EA 1039.74 USD InStock
10191-110
Oxalate (Oxalic Acid) Colorimetric Assay Kit, BioVision
Assays Protein Assays
Sensitive, High-Throughput adaptable
  • Simple, convenient and high throughput adaptable
  • Kit can detect Oxalate levels lower than 20 µM.
Oxalate (C2O42-), in the form of Oxalic acid is present in many foods and beverages (e.g. spinach, tea etc.). It accumulates in many plant tissues and play role in regulating pH, osmosis and calcium storage. In animals, oxalate is either absorbed from dietary intake or produced from glycolate metabolism in liver. Under normal conditions, the daily oxalate load can be excreted by kidney. However, hereditary defects can cause an increased level of oxalate, which leads to hyperoxaluria, and results in the formation of kidney stones. Therefore, measurement of oxalate level is useful for the prevention, diagnosis and monitoring of kidney stones. BioVision’s Oxalate Assay kit is an easy-to-use, sensitive and high throughput adaptable kit. In this assay, Oxalate reacts with Oxalate Converter & Oxalate Enzyme Mix to form an intermediate, which in turn reacts with a highly specific probe to generate color at 450 nm. The assay kit can detect Oxalate levels lower than 20 µM.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty