Would you like to log in again?
Oxalate Decarboxylase Activity Colorimetric Assay Kit, BioVision
Supplier: BioVision
AB196993-100TEST
10191-126EA 1072.51 USD InStock
10191-126
Oxalate Decarboxylase Activity Colorimetric Assay Kit, BioVision
Assays Enzyme Assays
Only Kit on the market to measure Oxalate Decarboxylase Activity
  • Simple, convenient and high throughput adaptable
  • Kit can detect Oxalate decarboxylase activity less than 20 µU.
Oxalate Decarboxylase (OXDC, EC 4.1.1.2) belongs to the cupin superfamily and is composed of two β-barrel domains. OXDC catalyzes the conversion of Oxalate into Formate and CO2 and plays an important role in stress response. In humans, high levels of oxalate can lead to various health problems including hyperoxaluria, kidney stones, and renal failure. Wood rotting fungi generates high levels of oxalate, causing rot in many crops including lettuce, soybean, dry bean, and tomato etc. Recent studies show that overexpression of OXDC in plants (e.g. tomato, soybean, lettuce, and tobacco) results in transgenic plants with resistance to fungal pathogenesis. Accurate measurement of Oxalate Decarboxylase activity is useful for a variety of therapeutic, diagnostic, and mechanistic studies. BioVision’s Oxalate Decarboxylase Activity Assay kit provides a quick and easy way to measure Oxalate Decarboxylase activity in various samples. In this assay, Oxalate Decarboxylase converts oxalate to formate, which subsequently converts a nearly colorless probe to a colored product with strong absorbance at 450 nm. The assay is simple, sensitive, high-throughput adaptable and can detect less than 20 µU of oxalate decarboxylase activity in a variety of samples.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty