Would you like to log in again?
Lysozyme Activity Assay Kit (Fluorometric), BioVision Inc.
Supplier: BioVision
AB211113-100TEST
75832-916EA 904.55 USD InStock
75832-916
Lysozyme Activity Assay Kit (Fluorometric), BioVision Inc.
Assays Enzyme Assays
Highly Sensitive and validated using many sample types
  • Rapid, and convenient
  • The assay can detect Lysozyme activity as low as 2 µU/ml in a variety of biological samples
Lysozyme (EC 3.2.1.17), also known as muramidase or N-acetylmuramide glycanhydrolase, is a hydrolase acting on glycosidic bonds. It hydrolyzes the β-(1-4)-glucosidic linkage between N-acetyl-muraminic acid and N-acetyl-D-glucosamine residues present in the mucopolysaccharide cell wall. Lysozyme is ubiquitously found in a wide range of biological fluids such as tears, saliva and tissues. It exhibits antibacterial, antitumor and immune modulatory activities. Elevated concentrations of lysozyme in urine and serum have been reported in patients suffering leukemia, tuberculosis, megaloblastic anemias, acute bacterial infections, ulcerative colitis, severe renal insufficiency, pyelonephritis and nephritis. BioVision’s lysozyme activity assay kit utilizes the ability of lysozyme to cleave a synthetic substrate to release the free fluorophore which can be easily quantified (Ex/Em= 360/445 nm). This kit provides a simple, ultra-sensitive assay that can detect as low as 2 µU/ml of Lysozyme activity in a variety of biological samples.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty