Would you like to log in again?
COX-2 Inhibitor Screening Kit (Fluorometric), BioVision Inc.
Supplier: BioVision
AB283401-100TEST
76020-878EA 657.83 USD InStock
76020-878
COX-2 Inhibitor Screening Kit (Fluorometric), BioVision Inc.
Assays Enzyme Assays
Simple assay to screen/study/characterize COX-2 Inhibitors
  • Simple method to screen COX-2 inhibitors
  • High-throughput
  • Includes Inhibitor Control, Celecoxib
Cyclooxygenase (COX), also known as prostaglandin-endoperoxide synthase (PTGS, EC 1.14.99.1), is an enzyme that is responsible for the formation of important biological mediators called prostanoids, including prostaglandins, prostacyclin and thromboxane. COX is the central enzyme in the biosynthetic pathway to prostanoids from arachidonic acid. There are two known isoenzymes: COX-1 and COX-2. COX-1 is constitutively expressed in many tissues and is the predominant form in gastric mucosa and in kidney. COX-2 is not expressed under normal conditions in most cells, but elevated levels are found during inflammation. Pharmacological inhibition of COX by non-steroidal anti-inflammatory drugs (NSAID) can provide relief from the symptoms of inflammation and pain. BioVision’s COX-2 Inhibitor Screening Kit offers a rapid, simple, sensitive, and reliable test suitable for high-throughput screening of COX-2 inhibitors. The assay is based on the fluorometric detection of Prostaglandin G2, the intermediate product generated by the COX enzyme.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty