Would you like to log in again?
Cyclooxygenase (COX) Activity Assay Kit (Fluorometric), BioVision
Supplier: BioVision
AB204699-100TEST
76020-882EA 673.13 USD InStock
76020-882
Cyclooxygenase (COX) Activity Assay Kit (Fluorometric), BioVision
Assays Enzyme Assays
Measures specifically COX-1 and COX-2 activity
  • Simple, rapid & convenient assay to measure both COX-1 and COX-2 activity
  • High-throughput adaptable
  • Includes COX-1 inhibitor, SC560 & COX-2 Inhibitor, Celecoxib
Cyclooxygenase (COX), also known as prostaglandin-endoperoxide synthase (PTGS, EC 1.14.99.1), is an enzyme that is responsible for the formation of important biological mediators called prostanoids, including prostaglandins, prostacyclin and thromboxane. COX is the central enzyme in the biosynthetic pathway to prostanoids from arachidonic acid. There are two known isoenzymes: COX-1 and COX-2. COX-1 is constitutively expressed in many tissues and is the predominant form in gastric mucosa and in kidney. COX-2 is not expressed under normal conditions in most cells, but elevated levels are observed during inflammation. Pharmacological inhibition of COX by non-steroidal anti-inflammatory drugs (NSAID) can provide relief from the symptoms of inflammation and pain. BioVision’s COX Activity Assay Kit provides a simple, sensitive, and high-throughput adaptable method to detect the peroxidase activity of COX in biological samples or purified/crude enzyme preparations. The kit includes COX-1 and COX-2 specific inhibitors to differentiate the activity of COX-1 and COX-2 as well as other peroxidases, which may be present in the sample. Detection limit: 6 μU/mg.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty