Would you like to log in again?
Albumin (BCG) Assay Kit (Colorimetric) , BioVision
Supplier: BioVision
AB235628-100TEST
76020-888EA 491.21 USD InStock
76020-888
Albumin (BCG) Assay Kit (Colorimetric) , BioVision
Assays Protein Assays
Simple, Highly Sensitive Assay, HTS
  • Simple procedure, High-throughput suitable
  • The assay can detect as low as 5 µg (0.01 g/dl) of albumin (BCG) in serum samples.
Albumin is the most abundant protein in human blood and is highly conserved among vertebrates. It plays a pivotal physiological role in maintenance of plasma osmotic pressure, vascular permeability, and transport of cholesterol, bile pigments, nitric oxide, metals, and other small molecules in the body. It also functions as a free radical scavenger of reactive oxygen and nitrogen species, triggers cell signaling processes, possesses anti-inflammatory and coagulatory effects. BioVision’s Albumin (BCG) Assay Kit is a simple high-throughput assay that detects Albumin concentration in serum. The assay is based on the selective interaction between Bromocresol Green (BCG) and Albumin forming a chromophore that can be detected at 620 nm. The signal is directly proportional to the amount of Albumin present in the serum. BCG does not react with other abundant plasma proteins like IgG. The assay can detect as low as 5 µg (0.01 g/dl) of albumin in serum samples.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty