Would you like to log in again?
Cathepsin H Inhibitor Screening Kit (Fluorometric), BioVision
Supplier: BioVision
AB211080-100TEST
76022-138EA 1208.56 USD InStock
76022-138
Cathepsin H Inhibitor Screening Kit (Fluorometric), BioVision
Assays Enzyme Assays
Only kit on the market to screen Cathepsin H inhibitors
  • Convenient & Rapid
  • High-throughput adaptable assay to screen/study/characterize the potential inhibitors of Cathepsin H
  • Cathepsin H inhibitor is included in the kit
Cathepsin H (CTSH, EC 3.4.22.16), a lysosomal cysteine protease, is important in the overall degradation of lysosomal proteins. It is composed of a dimer of disulfide-linked heavy and light chains, both produced from a single protein precursor. The encoded protein, which belongs to the peptidase C1 protein family, can act both as an aminopeptidase and as an endopeptidase. Increased expression of this gene has been correlated with malignant progression of prostate tumors. BioVision’s Cathepsin H Inhibitor Screening Kit utilizes the ability of an active Cathepsin H to cleave a synthetic AFC-based peptide substrate to release free AFC, which can be easily quantified using a fluorometer or fluorescence microplate reader. In the presence of a Cathepsin H-specific inhibitor, the cleavage of this substrate is reduced/abolished resulting in decrease or total loss of the AFC fluorescence. This simple and high-throughput adaptable assay kit can be used to screen/study/characterize potential inhibitors of Cathepsin H.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty