Would you like to log in again?
Cathepsin E Inhibitor Screening Kit (Fluorometric), BioVision
Supplier: BioVision
AB211082-100TEST
76022-142EA 1616.44 USD InStock
76022-142
Cathepsin E Inhibitor Screening Kit (Fluorometric), BioVision
Assays Enzyme Assays
Only kit on the market to screen Cathepsin E inhibitors
  • Convenient & Rapid
  • High-throughput adaptable assay to screen/study/characterize the potential inhibitors of Cathepsin E
  • Cathepsin E inhibitor is included in the kit
Cathepsin E (CTSE, EC: 3.4.23.34) is a gastric aspartyl protease that functions as a disulfide-linked homodimer. This protease has a specificity similar to that of pepsin A and cathepsin D. It is an intracellular proteinase that is found in highest concentration on the surface of epithelial mucus-producing cells of the stomach. It is the first aspartic proteinase expressed in the fetal stomach and is found in more than half of the gastric cancers. BioVision’s Cathepsin E Inhibitor Screening Kit utilizes the ability of Cathepsin E to cleave a synthetic MCA-based peptide substrate to release free MCA, which can be easily quantified using a fluorometer or fluorescence microplate reader. In the presence of a Cathepsin E-specific inhibitor, the cleavage of the substrate is reduced/abolished resulting in decrease or total loss of the MCA fluorescence. This simple and high-throughput adaptable assay kit can be used to screen/study/characterize the potential inhibitors of Cathepsin E.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty