Would you like to log in again?
Intracellular Hydrogen Peroxide Detection Kit (Cell-Based), BioVision
Supplier: BioVision
AB239701-200TEST
76022-144EA 553.68 USD InStock
76022-144
Intracellular Hydrogen Peroxide Detection Kit (Cell-Based), BioVision
Assays Cellular Assays
Simple assay to screen stimulators/inhibitors that affect intracellular levels of hydrogen peroxide
  • Simple procedure
  • Histochemical method for identification of intracellular H2O2 in the samples
Hydrogen Peroxide (H2O2) is a reactive oxygen species and by-product of oxygen metabolism. H2O2 serves as a key regulator for a number of oxidative stress related pathways and is linked to asthma, inflammatory arthritis, atherosclerosis, diabetic vasculopathy, osteoporosis, and a number of neurodegenerative diseases. BioVision’s Intracellular Hydrogen Peroxide Detection kit uses a dye that reacts with intracellular hydrogen peroxide to produce an orange color and fluorescence, which is proportional to the concentration of intracellular hydrogen peroxide. This kit provides a simple, easy to perform, histochemical method for identification of intracellular H2O2 in the samples.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty