Would you like to log in again?
Microsome Isolation Kit, BioVision Inc.
Supplier: BioVision
AB206995-50TEST
76022-146EA 871.01 USD InStock
76022-146
Microsome Isolation Kit, BioVision Inc.
Assays Cellular Assays
Simple method to isolate microsomes
  • Simple & convenient protocol
  • The kit provides unique formulations of ready-to-use buffers and reagents to isolate microsomes without the need for ultracentrifugation or sucrose gradient fractionation
Microsomes are spherical vesicle-like structures formed from membrane fragments following homogenization and fractionation of eukaryotic cells. The microsomal subcellular fraction is prepared by differential centrifugation and consists primarily of membranes derived from the endoplasmic reticulum (ER) and Golgi apparatus. Microsomes isolated from liver tissue are used extensively in pharmaceutical development, toxicology and environmental science to study the metabolism of drugs, organic pollutants and other xenobiotic compounds by the cytochrome P450 monooxidase (CYP) enzyme superfamily. Microsomal preparations are an affordable and convenient in vitro system for assessing Phase I biotransformation reactions, as they contain all of the xenobiotic-metabolizing CYP isozymes and the membrane-bound flavoenzymes (such as NADPH P450-Reductase and cytochrome b5) required for function of the multicomponent P450 enzyme system. BioVision’s Microsome Isolation Kit enables preparation of active microsomes in about one hour, without the need for ultracentrifugation or sucrose gradient fractionation. The kit contains sufficient reagents for 50 isolation procedures, yielding microsomes from roughly 25 grams of tissue or cultured cells.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty