Would you like to log in again?
Thrombin Activity Fluorometric Assay Kit, BioVision
Supplier: BioVision
AB197006-100TEST
76022-156EA 1230.85 USD InStock
76022-156
Thrombin Activity Fluorometric Assay Kit, BioVision
Assays Protein Assays
This assay kit is simple, rapid and can detect Thrombin activity as low as 1 ng in samples
  • Simple & convenient protocol.
  • Can detect thrombin activity as low as .
  • Kit contains all necessary reagents for measuring thrombin activity.
Thrombin enzyme (Factor IIa, EC 3.4.21.5), a Serine protease, is an important clotting factor in the coagulation cascade that involves the conversion of soluble fibrinogen to insoluble active fibrin strands. In this pathway, Prothrombin is proteolytically converted into an active Thrombin. Thrombin is also a potent vasoconstrictor and mitogen implicated as a major factor in vasospasm following subarachnoid hemorrhage. Ruptured cerebral aneurysm blood clots around a cerebral artery releases thrombin, which in turn induces acute and prolonged narrowing of the blood vessel, potentially resulting in cerebral ischemia and infarction (stroke). In addition, it is a pro-inflammatory enzyme that may influence the onset and progression of atherosclerosis. Biovision's Thrombin activity assay kit utilizes the ability of Thrombin to proteolytically cleave a synthetic substrate and release a fluorophore, AMC, which can be easily quantified by fluorescence reader. This assay kit is simple, rapid and can detect Thrombin activity as low as 1 ng in samples.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty