Would you like to log in again?
Thrombin Cleavage Kit, BioVision
Supplier: BioVision
AB207000-500TEST
76022-160EA 1018.67 USD InStock
76022-160
Thrombin Cleavage Kit, BioVision
Assays Protein Assays
Robust and efficient cleavage of tags from target proteins containing Thrombin recognition sequence
  • Simple procedure, fast and efficient cleavage of tags
  • The kit contains an active Thrombin enzyme sufficient to cleave up to 5 mg of target protein.
  • A 6x His-tagged protein containing the Thrombin cleavage site is included as a control. Following cleavage of the target protein, Thrombin can be conveniently removed using a Heparin Sepharose column.
Thrombin enzyme (Factor IIa, EC 3.4.21.5) is an important clotting factor that controls the transformation of soluble fibrinogen to insoluble active fibrin strands. It is also used as a valuable biochemical tool due to its high proteolytic specificity. Thrombin cleavage site (e.g. Leu-Val-Pro-Arg-ll-Gly-Ser; ll: cleavage site) is widely incorporated within the linker region of fusion or affinity tagged recombinant proteins. After successful cleavage with Thrombin, affinity tags or fused proteins can be separated from the target protein. Biovision's Thrombin Cleavage Kit provides an easy approach to test and optimize cleavage conditions of a target fusion or affinity-tagged protein containing Thrombin-specific cleavage site. The kit contains an active Thrombin enzyme sufficient to cleave up to 5 mg of the target protein. A 6xHis-tagged protein containing the Thrombin cleavage site is included as a Cleavage Control Protein. Following cleavage of the target protein, Thrombin can be removed by passing the reaction mix through the Heparin Sepharose column (Catalog # 6553 and 6554).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty