Would you like to log in again?
Sphingomyelinase Activity Fluorometric Assay Kit, BioVision
Supplier: BioVision
AB287874-96TEST
76022-162EA 658.85 USD InStock
76022-162
Sphingomyelinase Activity Fluorometric Assay Kit, BioVision
Assays Enzyme Assays
Simple assay to measure sphingomyelinase activity
  • Simple, rapid & convenient
  • The assay can detect sphingomyleinase activity as low as 10 µU/ml in a variety of samples.
Sphingomyelinase (SMase) cleaves sphingomyelin to produce phosphocholine and ceramide. The activation of SMase leads to increased production of ceramide, which acts as a lipid second messenger that induces a variety of cell regulatory phenomena such as apoptosis, cell differentiation, cell proliferation and sterol homeostasis. BioVision’s SMase Fluorometric Assay Kit provides a simple and sensitive method for measuring neutral-SMase enzymatic activity using fluorescence (Ex/Em = 535/587 nm). Neutral sphingomyelinase (N-SMase) is a Mg2+ sensitive enzyme that can be activated by a host of physiologically relevant and structurally diverse molecules. In this assay, SMase converts sphingomyelin substrate to phosphocholine and ceramide, which is then further utilized to produce fluorescence. This high-throughput adaptable assay kit can detect sphingomyelinase activity as low as 10 µU/ml in a variety of samples.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty