Would you like to log in again?
Hepatic Steatosis Assay Kit, BioVision Inc.
Supplier: BioVision
AB284556-100TEST
76022-166EA 891.69 USD InStock
76022-166
Hepatic Steatosis Assay Kit, BioVision Inc.
Assays Cellular Assays
Dual method to study Hepatic Steatosis: Quantifies Triglycerides and visualizes lipid droplets
  • Only Kit on the market to use a dual method to study Hepatic Steatosis: Quantifies Triglycerides (colorimetric/fluorometric) and visualizes lipid droplets by staining with Oil Red O. The kit provides enough reagents for 100 assays for triglyceride quantitation and staining reagents enough for two 96-well plates, two 6-well plates, or four 100 mm culture dishes. Easy and simple protocol. This kit can be used to screen and evaluate steatosis risk of drug candidates and to study the mechanism of steatosis development. Chloroquine is included as a positive control for inducing steatosis in liver cells, such as HepG2.
Hepatic Steatosis, also known as fatty liver, is a disease characterized by an excessive accumulation of lipid droplets within the liver. This can be caused by multiple factors, including excessive alcohol intake, obesity, and other diseases with abnormal fat metabolism. Hepatic Steatosis is commonly used as a marker to determine the hepatotoxicity of drug candidates. BioVision’s Hepatic Steatosis Kit provides reagents for measurement of intracellular triglyceride accumulation and staining reagents to visualize the lipid droplet formation in hepatocytes. The kit can be used to screen and evaluate steatosis risk of drug candidates and to study the mechanism of steatosis development. Chloroquine is included as a positive control for inducing steatosis in liver cells, such as HepG2. The kit provides enough reagents for 100 assays for triglyceride quantitation and staining reagents enough for two 96-well plates, two 6-well plates, or four 100 mm culture dishes.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty