Would you like to log in again?
CETP Inhibitor Screening Kit (Fluorometric) II, BioVision
Supplier: BioVision
AB283403-100TEST
76022-168EA 1011.22 USD InStock
76022-168
CETP Inhibitor Screening Kit (Fluorometric) II, BioVision
Assays Enzyme Assays
The most advanced kit on the market - includes human CETP enzyme and CETP inhibitor Anacetrapib
  • The most advanced kit on the market - includes human CETP enzyme and CETP inhibitor Anacetrapib for ultimate user-convenience. Simple and sensitive assay High-Throughput adaptable
Cholesteryl ester transfer protein (CETP) is a plasma protein that transfers a cholesteryl ester from HDL to LDL or VLDL in exchange for a triglyceride. HDL plays an important role in lipid metabolism and cardiovascular health. HDL transports cholesterol to the liver for excretion or to steroidogenic tissues for steroid synthesis. HDL also plays an important role in the reverse cholesterol transport pathway, removing cholesterol from lipid-filled macrophages, protecting against atherosclerosis. Because of this function, CETP is viewed as a target to increase HDL, with CETP inhibition being an active area of research and several CETP inhibitors at various stages of drug development. BioVision’s CETP Inhibitor Screening Kit uses a self-quenched fluorescent neutral lipid that can be measured when transferred to an acceptor molecule. The fluorometric intensity is directly proportional to the amount of neutral lipid transfer. Enriched Human CETP from plasma is included for screening. A CETP inhibitor (Anacetrapib) is included as a positive control.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty