Would you like to log in again?
PicoProbe™ LDH-Cytotoxicity Fluorometric Assay Kit, BioVision
Supplier: BioVision
AB197004-500TEST
76022-176EA 794.16 USD InStock
76022-176
PicoProbe™ LDH-Cytotoxicity Fluorometric Assay Kit, BioVision
Assays Cellular Assays
Ultra-sensitive Assay, HTS
  • Highly-Sensitive: Detects as less as 100 cells.
  • Rapid protocol: requires less than 20 min.
  • High-throughput adaptable.
Lactate Dehydrogenase (LDH, EC 1.1.1.27) is a stable enzyme and is present in all cell types. Cell damage leads to release of LDH enzyme into the media and its activity is widely used as a marker for cytotoxicity. It reversibly converts lactate into pyruvate, with the concomitant interconversion of NADH and NAD+. BioVision’s PicoProbe™ LDH-Cytotoxicity Fluorometric Assay Kit provides a sensitive, quick, and easy way for detection of LDH released from damaged cells. In this assay, LDH converts lactate to pyruvate and NADH, which reduces a proprietary probe to an intensely fluorescent product (Ex/Em = 535/587 nm). The amount of fluorescence is directly proportional to the number of damaged cells. The assay is adaptable to high-throughput format and can be completed in less than 20 min. Sensitivity: ~ 100 cells.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty