Would you like to log in again?
Cell-Mediated Cytotoxicity Fluorometric Assay Kit (7-AAD/CFSE), BioVision
Supplier: BioVision
AB287870-100TEST
76022-178EA 460.48 USD InStock
76022-178
Cell-Mediated Cytotoxicity Fluorometric Assay Kit (7-AAD/CFSE), BioVision
Assays Cellular Assays
Direct assay to measure cell-mediated cytotoxicity,
  • Direct method to measure cell-mediated cytotoxicity.
  • Does not require cell lysis.
  • Provides multi-parametric analysis.
Cell-mediated cytotoxicity is an important phenomenon characterized by cytolysis of a compromised cell in the body by immune system. Activation of immune system leads to removal of target cells infected by pathogens or transformed cells/cancer cells. This process is mediated by antibody-dependent cell-mediated cytotoxicity (ADCC), complement-mediated cytotoxicity, or lymphocyte-mediated cytotoxicity. BioVision’s Cell-Mediated Cytotoxicity Assay Kit contains carboxyfluorosuccinimide ester (CFSE), a green fluorescent probe that labels live target cells and 7-aminoactinomycin D (7-AAD), a red fluorescent probe that labels late apoptotic and necrotic target cells killed in the cytotoxicity assay. This assay does not require cell lysis and provides a direct measurement of cytotoxicity instead of indirect indicators such as release of ATP or lactate dehydrogenase activity. The flow cytometry-based method provides robust data and enables multi-parametric analysis.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty