Would you like to log in again?
Adenosine Deaminase Activity Assay Kit (Colorimetric), BioVision Inc.
Supplier: BioVision
AB211093-100TEST
76022-182EA 658.84 USD InStock
76022-182
Adenosine Deaminase Activity Assay Kit (Colorimetric), BioVision Inc.
Assays Enzyme Assays
Rapid & simple assay to measures total adenosine deaminase activity
  • Rapid, and convenient
  • The kit measures total Activity of Adenosine Deaminase with limit of quantification 1 mU recombinant Adenosine Deaminase.
Adenosine Deaminase (ADA) (E.C. 3.5.4.4.) is an enzyme that catalyzes the conversion of adenosine and 2’-deoxyadenosine to inosine and 2’-deoxyinosine. Adenosine Deaminase is widely distributed in various tissues and cells. There are two isoforms, ADA1 and ADA2. ADA1 is widely expressed in most cells in the body, particularly in lymphocytes and macrophages. It is present in the cytosol, nucleus and found associated with dipeptidyl peptidase-4 on the cell membrane. ADA2 was first found in the spleen but is predominantly found in the plasma and serum. Increased serum ADA levels are found in certain infectious diseases such as tuberculosis and various liver diseases such as acute hepatitis, alcoholic hepatic fibrosis, chronic active hepatitis to name a few. Adenosine Deaminase is also a marker for T-lymphocyte proliferation. In BioVision’s ADA Activity Assay, inosine formed from the breakdown of adenosine is converted to uric acid with ADA Convertor and ADA Developer. The uric acid is measured at OD 293 nm. The kit measures total Activity of Adenosine Deaminase with limit of quantification of 1 mU recombinant Adenosine Deaminase.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty