Would you like to log in again?
PicoProbe™ NADH Fluorometric Assay Kit, BioVision
Supplier: BioVision
AB234049-100TEST
76022-192EA 1067.96 USD InStock
76022-192
PicoProbe™ NADH Fluorometric Assay Kit, BioVision
Assays Enzyme Assays
The most sensitive kit on the market
  • Highly sensitive method to detect low levels of NADH.
  • Simple, fast and convenient.
  • This assay can measure less than 8 nM NADH in a variety of samples.
NAD and NADH exist in all living organisms and are important for energy transformation and maintenance of the redox state of cells or tissues. The reduced form, NADH is a driving force for cells and plays a critical role in cell regulation and repair processes. It is of increasing interest to be able to measure low level of NADH in samples or in enzymatic reactions. BioVision’s PicoProbe™ NADH Assay kit provides a highly sensitive method to detect low level of NADH (NAD is not detected in the reaction). There is no requirement to purify NADH from samples. The NADH Recycling Enzyme Mix specifically recognizes NADH (not NADPH) in the enzyme recycling reaction. The assay is rapid, simple and can measure less than 8 nM NADH in a variety of samples.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty