Would you like to log in again?
p300 Inhibitor Screening Assay Kit (Fluorometric), BioVision
Supplier: BioVision
AB196996-100TEST
76022-196EA 1118.49 USD InStock
76022-196
p300 Inhibitor Screening Assay Kit (Fluorometric), BioVision
Assays Enzyme Assays
Simple method to screen p300 Inhibitors
  • Simple procedure; takes ~ 45 min
  • Fast and convenient
  • Kit contains the necessary reagents for testing putative p300 inhibitors
p300 (EC 2.3.1.32) or KAT3B, a histone acetyltransferase, contributes to transcriptional activation by acetylating chromatin on the lysine residues of H3 and H4 histones. There is growing evidence that p300 plays an important role in cancer cell proliferation and differentiation. p300 inhibitors have potential applications in cancer therapy. BioVision’s p300 Inhibitor Screening Kit utilizes a H3 peptide and Acetyl CoA as the substrates. p300 acetylates the peptide and generates CoA-SH with a free thiol group. The CoA is detected using a Thiol Detecting Probe that reacts with thiol groups and gives enhanced fluorescence that can be measured at Ex/Em = 392/482 nm. In the presence of p300 specific inhibitors, the enzymatic activity is inhibited resulting in decreased or total loss of fluorescence. This assay kit is a simple, sensitive, and rapid tool to screen potential inhibitors of p300.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty